Learn More
Thermo Scientific™ Armadillo 96-well Clear PCR Plates, White Wells - PROMO
Product Code. 15361702
Click to view available options
Color:
Clear
Unit Size:
Pack of 25
Product Code. 15361702
Brand: Thermo Scientific™ AB3596
Only null left
Add to basket
Add to basket
The ultimate in high performance PCR plastics combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping.
  • Optically clear skirt—Aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design— Improved well surface area improves seal integrity in automated heat sealing
  • ANSI:SBS footprint—With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high throughput applications
  • Thermostable polycarbonate semi-skirted frame design—Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled U-bottom wells—Optimal heat transfer and maximum sample recovery
  • Indented Alphanumeric Grid reference lettering—Integrated frame and well design for improved heat sealing
  • Available in both clear and white colored wells—Ensures the highest level of sensitivity for all PCR/qPCR reactions
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Color Clear
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
No. of Wells 96
For Use With (Application) qPCR/Real-Time PCR
Plate Compatibility ANSI footprint ensures complete compatibility with robotic systems for high throughput applications
Quantity 25 Plates
Rimmed Raised
Well Color White