Learn More
ARL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10232-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a small GTP-binding protein of the RAS superfamily which functions as an ADP-ribosylation factor (ARF). The encoded protein is one of a functionally distinct group of ARF-like genes.Specifications
| ARL2 | |
| Polyclonal | |
| Unconjugated | |
| ARL2 | |
| ADP ribosylation factor like 2, ARFL2, ARL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 402, 56327, 65142 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08697, P36404, Q9D0J4 | |
| ARL2 | |
| ARL2 Fusion Protein Ag0334 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |