Learn More
Sartorius arium™ Water System Cartridge Set
Product Code. 14861412 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
arium™ Pro/611 UF Systems
arium™ Pro/611 VF or UV Systems
arium™ Pro/Basic/611 DI Systems
arium™ Systems w/Tap Water Feed
Type:
Analytical Kit
Biological Kit
Elemental Kit
Universal Kit
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Unit Size
14861412 arium™ Pro/611 VF or UV Systems Analytical Kit Each
14881372 arium™ Pro/611 UF Systems Biological Kit Each
14841382 arium™ Pro/Basic/611 DI Systems Elemental Kit Each
11747474 arium™ Systems w/Tap Water Feed Universal Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
*Tap water cartridges should be used only after the feed water specifications have been checked and following consultation with the responsible Sartorius™ application specialist.
Product Code. 14861412 Supplier Sartorius Supplier No. H2OAPACK
  • ONLINE EXCLUSIVE PRICE
    12150.00 NOK
    10461.24 NOK / Pack of 1
    Save 1688.76 NOK
    13% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
*Tap water cartridges should be used only after the feed water specifications have been checked and following consultation with the responsible Sartorius™ application specialist.
Produce ultrapure water beyond the ASTM Type 1 standard. Sartorius arium™ Water System Cartridge Set is designed for installation in arium™ Pro, Basic or 611 systems. Each consumable kit includes a pre-treatment and polishing cartridge. Kits are specifically formulated for each particular system.
arium™ water purification systems (Pro, Basic and/or 611 series) are designed to produce the highest purity laboratory water in the industry, with specifications that exceed the ASTM Type 1 standard. The cartridge sets are constructed of the highest quality materials with an advanced design to produce consistent, reproducible results.
  • Exceptional quality of the purification media
    • Optimized to remove organic and inorganic contaminants
    • Highly porous activated carbon combined with efficient ion exchange resins
    • Produces water with the lowest total organic carbon (TOC) content for systems using ultraviolet (UV) technology
  • Specially engineered so that water flows from top to bottom
    • The flow though the ion exchange medium prevents separation of the mixed-bed resin
    • Allows full use of each cartridge with no wasted media
    • Ensures a long cartridge life with unparalleled performance
  • Two-cartridge sets:
    • Pretreatment cartridge
    • Polishing cartridge
  • Specifically formulated for particular arium™ systems:
    • Analytical Kit for VF or UV systems
    • Biological Kit for UF systems
    • Environmental Kit for DI systems
    • Universal Kit for systems with tap water feed*
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM type 1
Capacity To 18.2 MΩ x cm ultrapure water in eq: 1.25
Feedwater Source Pretreated
Material Polypropylene
Purification Method Ion Exchange, Activated Carbon
Resin Type Activated Carbon and Mixed Bed Ion Exchange Resin
For Use With (Application) Standard, cell culture and chromatographic
For Use With (Equipment) arium™ Pro/611 VF or UV Systems
Includes Pretreatment, polishing cartridge
Type Analytical Kit
Dimensions (L x W x H) 450 x 230 x 160 mm (WxDxH)