Learn More
Sartorius Arium™ Tubing Extension Set
Product Code. 17214233 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium Smart Station Ultrapure Water
Arium™ Smart Station Pure Water
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17214233 Arium Smart Station Ultrapure Water 1 unit
17204233 Arium™ Smart Station Pure Water 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17214233 Supplier Sartorius Supplier No. H2OATESUP
  • ONLINE EXCLUSIVE PRICE
    2700.00 NOK
    2361.56 NOK / Pack of 1
    Save 338.44 NOK
    12% Off
In Stock
Extension set (4 m) for Arium™ Smart Station, used for uncomplicated extension of connection between Arium™ Smart Station and connected water system
  • Extended tubing set used for Smart Station ultrapure and pure for uncomplicated extension of standard supplied tubing set
  • Operating range can be easily increased from 2 to 4 m
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium Smart Station Ultrapure Water
Type Tubing