missing translation for 'onlineSavingsMsg'
Learn More

Aripiprazole

Product Code. p-7077705 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15452192 1 g 1g
15462192 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15452192

Brand: Thermo Scientific Acros 457990010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 129722-12-9 | C23H27Cl2N3O2 | 448.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129722-12-9
Molecular Formula C23H27Cl2N3O2
Molecular Weight (g/mol) 448.39
InChI Key CEUORZQYGODEFX-UHFFFAOYSA-N
Synonym aripiprazole, abilify, abilitat, abilify discmelt, discmelt, aripiprazol, opc 31, aripiprazole usan, aripiprazolum, aripirazole
PubChem CID 60795
ChEBI CHEBI:31236
IUPAC Name 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES C1CC(=O)NC2=C1C=CC(=C2)OCCCCN3CCN(CC3)C4=C(C(=CC=C4)Cl)Cl

Specifications

Melting Point 149.0°C
Loss on Drying 0.3% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetic acid, insoluble in methanol and hexane
Formula Weight 448.39
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Aripiprazole
compliance-icons
Product Identifier
  • Aripiprazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Aripiprazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.