missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ARHGDIG (Human) Recombinant Protein

Human ARHGDIG full-length ORF ( AAH47699, 1 a.a. - 225 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000398-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141871

  • 4130.00 NOK / 10µg
Estimated Shipment: 04-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MLGLDACELGAQLLELLRLALCARVLLADKEGGPPAVDEVLDEAVPEYRAPGRKSLLEIRQLDPDDRSLAKYKRVLLGPLPPAVDPSLPNVQVTRLTLLSEQAPGPVVMDLTGDLAVLKDQVFVLKEGVDYRVKISFKVHREIVSGLKCLHHTYRRGLRVDKTVYMVGSYGPSAQEYEFVTPVEEAPRGALVRGPYLVVSLFTDDDRTHHLSWEWGLCICQDWKD
Specifications

Specifications

AAH47699
Rho GDP dissociation inhibitor (GDI) gamma
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARHGDIG
GST
398
Wheat germ expression system
10 μg
RHOGDI-3
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers