Learn More
ARF6 Monoclonal Antibody (6ARF01), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA511795
Product Code. 11374513
  • 4540.00 NOK / 500µL
Description
MA5-11795 targets ADP-ribosylation Factor 6 in WB and ICC/IF applications and shows reactivity with Human and Mouse samples. The MA5-11795 immunogen is aRF6 protein.
ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ARF6
Monoclonal
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P62330, P62331
ARF6
ARF6 protein
500 μL
Primary
Human, Mouse
Antibody
IgG1 κ
Immunocytochemistry, Western Blot
6ARF01
Unconjugated
ARF6
ADP ribosylation factor 6; ADP-ribosylation factor 6; AI788669; ARF6; AW496366; DKFZp564M0264
Mouse
Protein G
RUO
11845, 382
4° C