Learn More
Thermo Scientific™ Arctic Express™ Dual Storage Systems
Product Code. 10338253 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 L
28 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10338253 10 L Each
10699463 28 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10338253 Supplier Thermo Scientific™ Supplier No. CK50920
  • ONLINE EXCLUSIVE PRICE
    29480.00 NOK
    27960.08 NOK / Pack of 1
    Save 1519.92 NOK
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a 'can and cane' storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
TRUSTED_SUSTAINABILITY
Specifications
Type Manual Fill LN2 Dewar
Capacity (Metric) 10 L
No. of Doors 1
Model Arctic Express Dual 10
Static Holding Time 21 shipper/42 lab
Diameter (English) 10.2 in.
Diameter (English) Inside Neck 2 in.
Diameter (Metric) 25.9 cm
Diameter (Metric) Inside Neck 5.08 cm
Vial Capacity 150 x 2mL