Learn More
Karl Hecht™ Assistent™ AR-Glas™ Round Bottom Test Tubes
Product Code. p-8003465
Change view
Click to view available options
Diameter (Metric):
8.0 mm
9.5 mm
10.5 mm
11.6 mm
13.25 mm
14.5 mm
15.25 mm
17.5 mm
Length (Metric):
40 mm
70 mm
85 mm
100 mm
160 mm
180 mm
Unit Size:
Pack of 100
Pack of 200
Pack of 500
Product Code. Diameter (Metric) Length (Metric) unitSize
10393651 11.6 mm 100 mm Pack of 100
10343751 13.25 mm 100 mm Pack of 100
10043811 17.5 mm 100 mm Pack of 100
10109880 9.5 mm 100 mm Pack of 100
10185941 15.25 mm 160 mm Pack of 200
10262101 17.5 mm 180 mm Pack of 100
11381535 8.0 mm 40 mm Pack of 500
10706382 14.5 mm 85 mm Pack of 100
10664101 10.5 mm 70 mm Pack of 100
9 options
Product Code. 10393651
Brand: Karl Hecht™ 42775027
Only null left
Add to basket
Add to basket
Ideal for routine laboratory analysis and procedures. Karl Hecht™ AR-Glas™ Round Bottom Test Tubes are manufactured from durable AR-Glas soda glass.
  • Rimless
  • With heavy wall
  • Available in a range of sizes
Specifications
Length (Metric) 100 mm
Packaging 100Pack; Box
Rimmed No
Diameter (Metric) 11.6 mm
Product Type Test Tube