Learn More
Karl Hecht™ Assistent™ AR-Glas™ Round Bottom Test Tubes
Description
- Rimless
- With heavy wall
- Available in a range of sizes
Specifications
Specifications
| Length (Metric) | 40 mm |
| Packaging | 100Pack; Box |
| Rimmed | No |
| Diameter (Metric) | 8.0 mm |
| Product Type | Test Tube |