Learn More
Karl Hecht™ Assistent™ AR-Glas™ Round Bottom Test Tubes
Product Code. 10393651
Click to view available options
Diameter (Metric):
8.0 mm
9.5 mm
10.5 mm
11.6 mm
13.25 mm
14.5 mm
15.25 mm
17.5 mm
Length (Metric):
40 mm
70 mm
85 mm
100 mm
160 mm
180 mm
Unit Size:
Pack of 100
Pack of 200
Pack of 500
Product Code. 10393651
Brand: Karl Hecht™ 42775027
Only null left
Add to basket
Add to basket
Ideal for routine laboratory analysis and procedures. Karl Hecht™ AR-Glas™ Round Bottom Test Tubes are manufactured from durable AR-Glas soda glass.
  • Rimless
  • With heavy wall
  • Available in a range of sizes
Specifications
Length (Metric) 100 mm
Packaging 100Pack; Box
Rimmed No
Diameter (Metric) 11.6 mm
Product Type Test Tube