Learn More
BRAND™ AR-Glas™ Stirring Rods
Product Code. 10069920 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
200 mm
250 mm
Unit Size:
Pack of 50
Product Code. Length (Metric) unitSize
10069920 250 mm Pack of 50
10672242 200 mm Pack of 50
2 options
Product Code. 10069920
Brand: BRAND™ 135815
Only null left
Add to basket
Add to basket
Glass stirring rods
TRUSTED_SUSTAINABILITY
Specifications
Description Fused closely at both ends
Diameter (Metric) 6 mm
Length (Metric) 250 mm
Quantity 100