Learn More
Notrax Aqua Trap™ Robust Barrier Mat with Raised Domes
Product Code. p-7227233
Change view
Click to view available options
Length (English):
35 in.
59 in.
71 in.
Unit Size:
Each
Product Code. Length (English) unitSize
17761948 35 in. Each
17701958 59 in. Each
17741958 71 in. Each
3 options
Product Code. 17761948
Brand: Notrax 150S0023CH
Only null left
Add to basket
Add to basket
True to its name, this patented perimeter Aqua Dam forms a tray that can hold up to 6 liters of water per square meter and is treated with an anti microbial carpet treatment to stop bacteria and germs at the entrance.
  • True to its name, this patented perimeter ''Aqua Dam'' forms a tray that can hold up to 6 liters of water per square meter
  • The textile top surface is high pressure moulded into a bubble design on an indestructible rubber base to withstand the heaviest of foot traffic
  • Unsurpassed ability to scrape debris from shoes while maintaining its great looking appearance for the longest time
  • Unique slip resistant gripper pattern on the back
Specifications
Color Charcoal
Length (English) 35 in.
Length (Metric) 90 cm
Material Polyester
Type Industrial and Commercial Matting
Width (Metric) 60 cm