Learn More
3M™ Nomad™ Aqua Textile Drop Down Mat, Series 85
Product Code. 17108580
Click to view available options
Dimensions (L x W):
150 x 91.4 cm
200 x 130 cm
300 x 130 cm
300 x 200 cm
600 x 130 cm
Color:
Black
Gray
Unit Size:
Each
Product Code. 17108580
Brand: 3M™ Nomad™ 7000032562
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Advanced water removal and retention properties keep floors cleaner and safer, for an extended period of time. Traditional, deep cut pile and coir matting allows dirt and moisture to sit on the surface and be tracked into the building.
  • High performing textile entrance mat, with advanced water removal and retention properties
  • Stops 80% of dirt and moisture at the door
  • Entrance mat for high traffic applications (1500 to 5000 crossings per day)
  • Phthalate-free, slip-resistant PVC backing
  • Excellent appearance retention
  • Long lasting and easy to clean
  • Available as drop down mats or in rolls
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 300 x 200 cm
For Use With (Application) Heavy traffic areas
Model Nomad™ Aqua, Series 85
Resistance Slip-resistant
Thickness (Metric) 7.5 mm
Color Gray
Length (Metric) 200 cm
Material PVC (Backing)