Learn More
Applied Biosystems™ Total RNA Control (Human)
Product Code. 10618095 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10618095 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10618095 Supplier Applied Biosystems™ Supplier No. 4307281
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Provided at a convenient reverse-transcription-ready concentration for all RT-PCR reactions
TaqMan™ Control Total RNA is from Raji cells (Burkitt's lymphoma), and is conveniently packaged in a PCR-ready concentration for use as a control template in RT-PCR reactions. The Human total RNA is provided at a concentration of 50ng/μL.
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Format Frozen
Content And Storage 100 μl Total Human RNA (at 50 ng/μl). Store at -15 to -25°C.
Species Human
Shipping Condition Dry Ice
Concentration 50 ng/μL
Label or Dye Unlabeled
Quantity 100 μL
For Research Use Only. Not for use in diagnostic procedures.
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.