missing translation for 'onlineSavingsMsg'
Learn More

APPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19885-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889344

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.
Specifications

Specifications

APPL1
Polyclonal
Unconjugated
APPL1
APPL, APPL1, Dip13 alpha, DIP13A, DIP13alpha, KIAA1428
Rabbit
Antigen Affinity Chromatography
RUO
26060, 290537, 72993
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K3H0, Q9UKG1
Appl1
APPL1 Fusion Protein Ag13703
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers