missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19885-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.Specifications
APPL1 | |
Polyclonal | |
Unconjugated | |
APPL1 | |
APPL, APPL1, Dip13 alpha, DIP13A, DIP13alpha, KIAA1428 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26060, 290537, 72993 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K3H0, Q9UKG1 | |
Appl1 | |
APPL1 Fusion Protein Ag13703 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |