missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein H Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4D9A4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66074-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887615

  • 3468.15 NOK / 150µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
Specifications

Specifications

Apolipoprotein H
Monoclonal
0.78 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02749, P26644, Q01339
Apoh
Apolipoprotein H Fusion Protein Ag17915
150 μL
Primary
Rat, Porcine, Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
4D9A4
Unconjugated
APOH
Anticardiolipin cofactor, APC inhibitor, Apo H, APOH, Apolipoprotein H, B2G1, B2GPI, Beta 2 glycoprotein 1, Beta 2 glycoprotein I, Beta(2)GPI, BG
Mouse
Protein G
RUO
100522100, 11818, 287774, 350
-20°C
Liquid
Videos
SDS
Documents
Certificates
Special Offers

Special Offers