missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody recognizes the endogenous ApoL1 of 39-45 kDa in blood lysate. This antibody is specific to APOL1.
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein L gene family encodes six highly homologous proteins designated apoL-I to -VI, which are associated with large high density type lipoproteins (HDL). The human apoL family maps to chromosome 22q12.1- 13.1 within a 127,000-bp region. apoL has been characterized as a pancreas specific, 383-amino acid, 42 kDa protein that contains a 12-amino acid secretory signal peptide. The apoL genes have TATA-less promoters and contain putative sterol regulatory elements, suggesting that transcription of these genes may be coordinated with that of the low density lipoprotein receptor and genes in pathways involving the synthesis of triglycerides and cholesterol. apoL homologs can undergo 10 fold changes in expression during atherosclerotic changes in vascular endothelial cells, which includes the inflammatory reaction of atherosclerotic lesions.
Specifications
Specifications
| Antigen | APOL1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 1G12D11 |
| Concentration | 1.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | APOL1 |
| Gene Accession No. | O14791 |
| Gene Alias | APO L, APOL, APOL I, APOL1, Apolipoprotein L, Apolipoprotein L I, apolipoprotein L, 1, Apolipoprotein L1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?