missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ APOF (Human) Recombinant Protein
Human APOF full-length ORF ( AAH26257, 37 a.a. - 327 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000319-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: TSYGKQTNVLMHFPLSLESQTPSSDPLSCQFLHPKSLPGFSHMAPLPKFLVSLALRNDALEEAGCQADVWALQLQLYRQGGVNATQVLIQHLRGLQKGRSTERNVSVEALASALQLLAREQQSTGRVGRSLPTEDCENEKEQAVHNVVQLLPGVGTFYNLGTALYYATQNCLGKARERGRDGAIDLGYDLLMTMAGMSGGPMGLAISAALKPALRSGVQQLIQYYQDQKDANISQPETTKEGLRAISDVSDLEETTTLASFISEVVSSAPYWGWAIIKSYDLDPGAGSLEI
Specifications
AAH26257 | |
apolipoprotein F | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOF | |
GST |
319 | |
Wheat germ expression system | |
25 μg | |
Apo-F, DKFZp781G18150, LTIP, MGC22520 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |