missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APJ Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20341-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the G protein-coupled receptor gene family. The encoded protein is related to the angiotensin receptor, but is actually an apelin receptor that inhibits adenylate cyclase activity and plays a counter-regulatory role against the pressure action of angiotensin II by exerting hypertensive effect. It functions in the cardiovascular and central nervous systems, in glucose metabolism, in embryonic and tumor angiogenesis and as a human immunodeficiency virus (HIV-1) coreceptor. Two transcript variants resulting from alternative splicing have been identified.Specifications
APJ | |
Polyclonal | |
Unconjugated | |
APLNR | |
AGTRL1, Angiotensin receptor like 1, apelin receptor, APJ, APJR, APLNR, G protein coupled receptor APJ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
187, 23796 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35414, Q9WV08 | |
APLNR | |
APJ Fusion Protein Ag14138 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |