Learn More
Fisherbrand™ Apiezon Grease
Product Code. p-8012711 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 g
100 g
Product Type:
AP101 Grease
L Grease
M Grease
Unit Size:
100g
50g
Product Code. Quantity Product Type unitSize
12971141 100 g M Grease 100g
12961141 50 g L Grease 50g
12991141 50 g AP101 Grease 50g
3 options
Product Code. 12971141
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lubricate pumps with this grease. Fisherbrand™ Apiezon Grease is designed for use with Fisherbrand™ rotary vane pumps.
  • Apiezon grease
  • For general use in Fisherbrand rotary vane pumps
TRUSTED_SUSTAINABILITY
Specifications
Product Type M Grease
Pressure 20°C 1.7x10-9 torr
Quantity 100 g
Temperature (Metric) 10°C to 30°C