Learn More
Fisherbrand™ Apiezon Grease
Product Code. 12961141 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
50 g
100 g
Product Type:
AP101 Grease
L Grease
M Grease
Unit Size:
100g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Unit Size
12961141 50 g L Grease 50g
12971141 100 g M Grease 100g
12991141 50 g AP101 Grease 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12961141
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Lubricate pumps with this grease. Fisherbrand™ Apiezon Grease is designed for use with Fisherbrand™ rotary vane pumps.
  • Apiezon grease
  • For general use in Fisherbrand rotary vane pumps
TRUSTED_SUSTAINABILITY
Specifications
Product Type L Grease
Pressure 20°C 7x10-11 torr
Quantity 50 g
Temperature (Metric) 10°C to 30°C