Learn More
APEH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543138
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GSTGFGQDSI LSLPGNVGHQ DVKDVQFAVE QVLQEEHFDA SHVALMGGSH Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%
APEH Polyclonal antibody specifically detects APEH in Human samples. It is validated for Western Blot.Specifications
APEH | |
Polyclonal | |
Unconjugated | |
APEH | |
AARE; ACPH; Acylamino-acid-releasing enzyme; acylamino-acid-releasing enzyme-like; acylaminoacyl-peptidase; acylaminoacyl-peptide hydrolase; acylpeptide hydrolase; Acyl-peptide hydrolase; Apeh; APH; cb5; D3F15S2; D3S48E; DNF15S2; LOW QUALITY PROTEIN: acylamino-acid-releasing enzyme; acylamino-acid-releasing enzyme; MGC2178; N-acylaminoacyl peptide hydrolase; N-acylaminoacyl-peptide hydrolase; OPH; Oxidized protein hydrolase; sb:cb5; wu:fi37d02 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
327 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P13798 | |
APEH | |
Synthetic peptide directed towards the middle region of human APEH | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |