Learn More
Thermo Scientific™ ApaI (10 U/μL)
Product Code. p-4521530 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3000 U
5,000 units
Unit Size:
3000 units
5000 units
Product Code. Quantity unitSize
10130690 3000 U 3000 units
10550841 5,000 units 5000 units
2 options
Product Code. 10130690
Brand: Thermo Scientific™ ER1411
Edge
Cut at GGGCC^C sites with ApaI restriction enzyme, which functions best at 37°C in B buffer (Isoschizomers: PspOMI).
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications
  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP
Note
Incubation at 30°C results in a 2-fold increase in activity. ApaI is inhibited by salt concentrations above 50 mM. ApaI cleavage is impaired by overlapping dcm methylation. To avoid dcm methylation, use a dam-, dcm- strain, such as GM2163.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10 U/μL
Components 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
Incubator Temperature 37°C
Quantity 3000 U
Product Type Apal