Learn More
mb2 Polyurethane Stool on Glides - Round
Product Code. 15734308
Change view
Click to view available options
Standard Height, Gliders
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15734308 Standard Height, Gliders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15734308 Supplier mb2 Supplier No. AP6B1D1NE3
  • ONLINE EXCLUSIVE PRICE
    1810.00 NOK
    1789.91 NOK / Pack of 1
    Save 20.09 NOK
    1% Off
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Polyurethane stool on glides - Round - Standard lift - Fix mechanism
  • Stool in polyurethane, round
  • Seat in polyurethane
  • Pneumatic cylinder standard
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Glides
Specifications
Product Type Stool
Color Black
Adjustable (Height) Yes
Adjustable Height
Autoclavable Not Autoclavable
Frame Material Steel
Includes Five-star nylon and fiberglass base. Gliders.
Quantity 1 Ea.
Seat Material Polyurethane
Seating Height 410 to 530 mm
Warranty 5 Years
Weight (Metric) 5.40 kg
With Armrest No
With Backrest No
With Casters No
Footrest No
With Lumbar Support No
With Tilt No
Material Polyurethane
Seating Type Stool