Learn More
mb2 Polyurethane Stool on Castors - Round
Description
- Stool in polyurethane, round
- Seat in polyurethane
- Pneumatic cylinder standard
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- Hard floor castors 50 mm
Specifications
Specifications
| Product Type | Stool |
| Color | Black |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Autoclavable | Not Autoclavable |
| Frame Material | Steel |
| Includes | Five-star nylon and fiberglass base. Casters. |
| Quantity | 1 Ea. |
| Seat Material | Polyurethane |
| Seating Height | 430 to 550 mm |
| Show More |