Learn More
mb2 Polyurethane Stool on Glides - Round
Description
- Stool in polyurethane, round
- Seat in polyurethane
- Pneumatic cylinder high
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- Glides
Specifications
Specifications
| Product Type | Stool |
| Color | Black |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Autoclavable | Not Autoclavable |
| Frame Material | Steel |
| Includes | Five-star nylon and fiberglass base. Gliders. |
| Quantity | 1 Ea. |
| Seat Material | Polyurethane |
| Seating Height | 570 to 830 mm |
| Show More |