Learn More
AP5Z1/SPG48 Mouse, anti-Human, CoraLite 594, Clone: 1G1D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66533-100UL
Description
AP5Z1, also known as SPG48 and KIAA0415, is a part of AP-5. Biallelic mutations in the AP5Z1 gene encoding the AP5Z1 subunit have been described in a small number of patients with hereditary spastic paraplegia (HSP). AP5Z1 physically interacts with other HSP proteins and that patient cells are sensitive to DNA damaging drugs.
Specifications
| AP5Z1/SPG48 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Immunofluorescence | |
| 1G1D7 | |
| CoraLite 594 | |
| 100 μL | |
| 9907 | |
| Liquid |