missing translation for 'onlineSavingsMsg'
Learn More

AP2M1 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30499681 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499681 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499681 Supplier Novus Biologicals Supplier No. NBP335403AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AP2M1 Polyclonal antibody specifically detects AP2M1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2M1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Adaptin-mu2, adaptor-related protein complex 2, mu 1 subunit, AP-2 mu chain, Clathrin assembly protein complex 2 medium chain, clathrin coat adaptor protein AP50, Clathrin coat assembly protein AP50, Clathrin coat-associated protein AP50, clathrin-associated/assembly/adaptor protein, medium 1, HA2 50 kDa subunit, KIAA0109, mu subunit, Plasma membrane adaptor AP-2 50 kDa protein, plasma membrane adaptor AP-2 50kDA protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 134-433 of human AP2M1 (NP_004059.2).,, Sequence:, TQQGIKSQHQTKEEQSQITSQVTGQIGWRREGIKYRRNELFLDVLESVNLLMSPQGQVLSAHVSGRVVMKSYLSGMPECKFGMNDKIVIEKQGKGTADETSKSGKQSIAIDDCTFHQCVRLSKFDSERSISFIPPDGEFELMRYRTTKDIILPFRVIPLVREVGRTKLEVKVVIKSNFKPSLLAQKIEVRIPTPLNTSGVQVICMKGKAKYKASENAIVWKIKRMAGMKESQISAEIELLPTNDKKKWARPPISMNFEVPFAPSGLKVRYLKVFEPKLNYSDHDVIKWVRYIGRSGIYET
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1173
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.