Learn More
Invitrogen™ Anza™ T4 PNK Kit
Product Code. 15461935
Change view
Click to view available options
Quantity:
50 reactions
Unit Size:
1 set
Product Code. Quantity unitSize
15461935 50 reactions 1 set
1 options
Product Code. 15461935
Brand: Invitrogen™ IVGN2304
Includes
Anza™ T4 PNK Enzyme (Polynucleotide Kinase), Anza™ 10X T4 PNK Buffer,
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • Part of the Anza Restriction Enzyme Cloning System
  • Quick DNA phosphorylation in 15 minutes
  • Heat inactivated in 5 minutes
Certifications
CoA
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 50 reactions Anza T4 PNK Enzyme
0.5 mL Anza 10X T4 PNK Buffer
Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme Polynucleotide Kinase
Compatible Buffer Anza™ 1X Buffer, Anza™ 10X T4 PNK Buffer
Quantity 50 reactions
Product Type T4 PNK Kit
Product Line Anza