Learn More
Invitrogen™ Anza™ 8 XhoI
Product Code. 15565680
Click to view available options
Quantity:
20,000 units
4000 units
Unit Size:
1 set
Product Code. 15565680
Brand: Invitrogen™ IVGN0088
Includes
Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 mL Anza 8 XhoI
2 mL Anza Buffer (10X)
2 mL Anza Red Buffer (10X)
Store at -20°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Xho I
Compatible Buffer Anza™ 10X Red Buffer, Anza™ 10X Buffer
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 20,000 units
Product Type Restriction Enzyme Cloning System
Research Category Traditional Cloning
Product Line Anza