Learn More
Invitrogen™ Anza™ 5-Pack Starter Kit
Product Code. 15521620
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Product Code. Quantity unitSize
15521620 1 kit Each
1 options
Product Code. 15521620
Brand: Invitrogen™ IVGN3004
Includes
Anza™ 11 EcoRI, Anza™ 5 BamHI, Anza™ 1 NotI, Anza™ 16 HindIII, Anza™ 12 XbaI, Anza™ T4 DNA Ligase Master Mix, Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer,
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA substrates
  • Complete digestion in 15 min
  • Includes 5 common restriction enzymes
  • Complete with Anza T4 DNA Ligase Master Mix and Anza Alkaline Phosphatase
Certifications
CoA
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage • 20 μL each of Anza 11 EcoRI, Anza 5 BamHI, Anza 1 NotI, Anza 16 HindIII, Anza 12 XbaI
• 100 μL Anza T4 DNA Ligase Master Mix
• 20 μL Anza Alkaline Phosphatase
• 500 μL Anza Buffer (10X)
• 500 μL Anza Red Buffer (10X)
Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Methylation Sensitivity Methylation Sensitivities Varies by Enzyme
Enzyme HindIII, NotI, Alkaline Phosphatase, XbaI, T4 DNA Ligase, BamHI, EcoRI
Compatible Buffer Anza™ 10X Red Buffer, Anza™ 10X Buffer
Type IIS RE No
Quantity 1 kit
Product Type Starter Kit
Research Category Traditional Cloning
Product Line Anza