Learn More
Invitrogen™ Anza™ 105 Hin1I
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN1056
Additional Details : Weight : 0.01000kg
Product Code. 15611972
  • 934.00 NOK / 1500 units
Estimated Shipment: 22-05-2024
to see stock.
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA types
  • Complete digestion in 15 min
  • Overnight digestion without star activity
Certifications
CoA
Specifications
Specifications
75 μL Anza 105 Hin1I
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)
Store at -20°C.
CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
Restriction Enzyme Cloning System
Anza™
Approved for shipment on Wet or Dry Ice
Hin1I
Yes
No
Traditional Cloning