missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ANXA8 (Human) Recombinant Protein
Human ANXA8 full-length ORF ( AAH04376, 1 a.a. - 327 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000244-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAWWKSWIEQEGVTVKSSSHFNPDPDAETLYKAMKGIGTNEQAIIDVLTKRSNTQRQQIAKSFKAQFGKDLTETLKSELSGKFERLIVALMYPPYRYEAKELHDAMKGLGTKEGVIIEILASRTKNQLREIMKAYEEDYGSSLEEDIQADTSGYLERILVCLLQGSRDDVSSFVDPGLALQDAQDLYAAGEKIRGTDEMKFITILCTRSATHLLRVFEEYEKIANKSIEDSIKSETHGSLEEAMLTVVKCTQNLHSYFAERLYYAMKGAGTRDGTLIRNIVSRSEIDLNLIKCHFKKMYGKTLSSMIMEDTSGDYKNALLSLVGSDP
Specifications
AAH04376 | |
annexin A8-like 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANXA8L2 | |
GST |
244 | |
Wheat germ expression system | |
25 μg | |
ANXA8, FLJ32754, FLJ54151, bA145E20.2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |