Learn More
Tork Antimicrobial Liquid Soap Refill for S4 System
Product Code. 30243757 Shop All Tork Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243757 Clear Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30243757 Supplier Tork Supplier No. 424709
  • ONLINE EXCLUSIVE PRICE
    1075.00 NOK
    1005.09 NOK / Pack of 6
    Save 69.91 NOK
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
Perfect for food-handling environments, Tork antimicrobial hand washing liquid soap based on lactic acid is tough on problem microorganisms.
  • Help to protect guests - this liquid soap conforms with EN1499 and EN14476 and is effective against all enveloped viruses.
  • This biodegradable liquid soap contains a 100% plant-based active ingredient.
  • Our hand soap is dermatologically tested and suitable for frequent use.
  • Meet safety standards - this non-flammable soap has no CLP symbols, warning phrases or transport restrictions.
  • 100% plant-based active ingredient
  • 100% biodegradable
  • Suitable for all Tork Skincare Dispensers (S4).
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Form Liquid
Certifications/Compliance EN1499, EN14476
Certifications/Compliance (Europe) EN1499, EN14476
Disposable Limited-use
Packaging Plastic Bottle
Quantity 6/Pk.
Volume (Metric) 1 L
Weight (Metric) 6.719 kg