Learn More
Tork Antimicrobial Foam Soap Refill S4
Product Code. 16679921 Shop All Tork Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16679921 White Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16679921 Supplier Tork Supplier No. 520800
  • ONLINE EXCLUSIVE PRICE
    1459.05 NOK
    1348.14 NOK / Pack of 6
    Save 110.91 NOK
    7% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Ensure effective use against a variety of problematic organisms.
  • Ensure effective use against a variety of problematic organisms - our antimicrobial soap refill conforms with EN 1499.
  • Reduce the risk of cross-contamination - our sealed refills come with a single-use pump for improved hygiene.
  • Intuitive refilling in less than 10 seconds means effortless maintenance for you and your staff.
  • Lower your eco-footprint and reduce waste with our recyclable bottle that collapses as the soap is used.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Form Foam
Certifications/Compliance EN1499
Certifications/Compliance (Europe) EN1499
Disposable Limited-use
Packaging Plastic Bottle
Quantity 6/Pk.
Volume (Metric) 1 L