Learn More
Contec™ Anticon™ Gold HeavyWeight™ Wipes
BIOPHARMA
Product Code. 16338147 Shop All Contec Products
Change view
Click to view available options
Packaging Quantity:
150/Bag, 8 Bags/Cs.
2 Bags x 75 Wipes, 8 Bags/Cs.
Unit Size:
Pack of 1200
Product Code. Packaging Quantity unitSize
16338147 150/Bag, 8 Bags/Cs. Pack of 1200
10470225 2 Bags x 75 Wipes, 8 Bags/Cs. Pack of 1200
2 options
Product Code. 16338147
Brand: Contec™ GDHW0101
Only null left
Add to basket
Add to basket
Manufactured from 100% continuous filament polyester with a double-knit no-run interlock construction. Contec™ Anticon™ Gold HeavyWeight™ Wipes have sealed edges to reduce particle and fiber generation.
Anticon Gold HeavyWeight uses patented textile technology to achieve superior sorption characteristics for critical applications. The proprietary sealed edges of these wipes allows the use of 100% of the wipe for cleaning without the worry of harming delicate surfaces. The sealed edges also reduce particle and fiber generation. Anticon Gold HeavyWeight wipes' low sub-micron particles and fiber counts are ideal for use in the most critical environments. Cleanroom laundered and packaged. Also available with Particle Attraction Technology (P.A.T.).
Features & Benefits:
  • Recommended for ISO Class 3-8 environments
  • Sealed edges to reduce particle and fiber generation
  • Low sub-micron particles and fiber counts
  • High sorptive capacity and fast adsorption rate for superior spill control
  • High tensile strength to eliminate tearing and snagging in tough cleaning applications
  • Manufactured from 100% continuous filament polyester and specially processed for superior cleanliness and sorbency
  • Cleanroom laundered
  • Tested in accordance with the IEST Recommend Practices for the Evaluation of Wiping Materials for Critical Environments
  • Offered in bulk and stacked packaging
  • Available with Particle Attraction Technology (P.A.T.)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Wipe
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Polyester
Dimensions (L x W) 23 x 23 cm (9 x 9 in.)
Packaging Quantity 150/Bag, 8 Bags/Cs.