missing translation for 'onlineSavingsMsg'
Learn More

Antibiotic-5d, MedChemExpress

Product Code. 16443188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16443188 10 mg 10mg
16474466 100 mg 100mg
16465917 5 mg 5mg
16459736 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16443188

Brand: Medchem Express HY100833

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Antibiotic-5d is a synthesis and antimicrobial compound.

Antibiotic-5d is a synthesis and antimicrobial compound.

Specifications

Chemical Name or Material Antibiotic-5d
Physical Form Solid
CAS 251349-54-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H18N2O4S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (167.58 mM; Need ultrasonic)
SMILES O=C(C1=CSC([C@H]2N(C(OC(C)(C)C)=O)CCC2)=N1)O
Molecular Weight (g/mol) 298.36
Formula Weight 298.36
Percent Purity 98.35%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.