Learn More
anti-Von Willebrand Factor, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NB600-586-0.1ml
Encompass
Product Code. 15735527
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Von Willebrand Factor Polyclonal specifically detects Von Willebrand Factor in Human, Mouse, Canine, Feline samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC.