Learn More
VDR Monoclonal Antibody (9A7), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA1710
Additional Details : Weight : 1.24740kg
Description
MA1-710 detects vitamin D receptor (VDR) from human, rat, mouse, hamster, chicken, amphibian, and fish tissues. This antibody does not cross-react with estrogen or glucocorticoid receptors. MA1-710 detects both the occupied and unoccupied forms of the receptor. MA1-710 has been successfully used in Western blot, IF, ICC, IP, ChIP, and gel shift procedures. By Western blot, this antibody detects a 58 kDa and 60 kDa protein representing VDR in avians while the VDRs of mammalian species appear as single bands within the 52-55 kDa range. The MA1-710 immunogen is partially purified chicken intestinal cytoplasmic 1,25-(OH)2D3 receptor protein. This antibody recognizes an epitope between amino acid residues 89 and 105 of human VDR. This region is just C-terminal to the DNA binding domain of the VDR.
VDR Monoclonal antibody specifically detects VDR in Chicken, Human, Many, Mouse, Rat samples. It is validated for ChIP Assay, Gel Shift, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
VDR | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O42392, P11473, P13053, P48281 | |
VDR | |
Partially purified chicken intestinal cytoplasmic 1,25-(OH)2D3 receptor protein. | |
100 μg | |
Primary | |
Chicken, Human, Multi-species, Mouse, Rat | |
Antibody | |
IgG2b |
ChIP Assay, Gel Shift, Immunocytochemistry, Immunoprecipitation, Western Blot | |
9A7 | |
Unconjugated | |
VDR | |
1,25-dihydroxyvitamin D3 receptor; HGNC:12679; I79_025795; Nr1i1; Nuclear receptor subfamily 1 group I member 1; PPP1R163; protein phosphatase 1, regulatory subunit 163; vdr; vdr.S; vdr-A; vitamin D (1,25- dihydroxyvitamin D3) receptor; vitamin D (1,25- dihydroxyvitamin D3) receptor S homeolog; vitamin D (1,25-dihydroxyvitamin D3) receptor; vitamin D nuclear receptor variant 1; vitamin D receptor; vitamin D receptor protein; vitamin D receptor S homeolog; Vitamin D3 receptor; XELAEV_18015993mg; xVDR | |
Rat | |
Protein G | |
RUO | |
22337, 24873, 395988, 7421 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |