Learn More
Vimentin Chicken anti-Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine, Polyclonal, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 44 publications
Brand: Novus Biologicals NB300-223
Description
Vimentin Polyclonal specifically detects Vimentin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Vimentin | |
| Polyclonal | |
| Western Blot 1:10000-1:20000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:5000, Immunohistochemistry-Paraffin 1:150, Immunohistochemistry-Frozen | |
| P08670 | |
| VIM | |
| Full length recombinant human Vimentin expressed in and purified from E. coli. [Swiss-Prot# P08670] | |
| IgY purified | |
| RUO | |
| 7431 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Supplied as concentrated IgY in PBS with 0.02% Sodium Azide | |
| FLJ36605, vimentin | |
| Chicken | |
| 50 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine | |
| Precipitation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only