Learn More
Vancomycin Mouse anti-Chemical, Clone: 2F10, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125796
Description
MA1-25796 detects Vancomycin. MA1-25796 has been successfully used in immunofluorescence procedures. The MA1-25796 immunogen is Vancomycin. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Vancomycin is a branched tricyclic glycosylated peptide with bactericidal activity against most organisms and bacteriostatic effect on enterococci. At a site different from that of penicillins and cephalosporins, Vancomycin binds tightly to the D-alanyl-D-alanine portion of cell wall precursors, thereby interfering with bacterial cell wall synthesis. This leads to activation of bacterial autolysins that destroy the cell wall by lysis. Vancomycin may also alter the permeability of bacterial cytoplasmic membranes and may selectively inhibit RNA synthesis. [National Center for Biotechnology Information. PubChem Compound Database; CID=14969]
Specifications
| Vancomycin | |
| Monoclonal | |
| 2.9 mg/mL | |
| Vancocin; Vancoled; Vancomicina; Vancomycine; Vancomycinum; C66H75Cl2N9O24 | |
| Vancomycin. | |
| 250 μg | |
| Primary | |
| Antibody | |
| IgM |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 2F10 | |
| Unconjugated | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.