Learn More
Sartorius™ Anti-static Decontamination Wipes
Product Code. 10762207 Shop All Sartorius Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
:
Anti-static Decontamination Wipes
Product Code.
10762207 Anti-static Decontamination Wipes
1 missing translation for 'options'
Product Code. 10762207
missing translation for 'mfr': Sartorius™ YWCG07
Only null left
Add to basket
Add to basket
Decontamination Wipes are anti-static and used for cleaning the fan and filters on the Sartorius SWC Safety Weighing Cabinets, ensuring the units are left clean for the next analyst to use without the risk of contamination.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Anti-static Decontamination Wipe
For Use With (Equipment) Sartorius SWC Safety Weighing Cabinets