Learn More
3M™ Anti-Scratch/Anti-Fog Safety Glasses
Click to view available options
Lens Tint:
Amber
Clear
Gray
Shade 5
Unit Size:
Each
Pack of 20
Description
- Very lightweight, sleek and modern design safety glasses offering excellent coverage and field of vision
- Strong polycarbonate lens for impact resistance
- Optical class 1 for maximum clarity
- Inclinable lens for easy adjustment and maximum comfort
- Soft, flexible temples for wearer comfort
- Anti-Scratch and Anti-Fog coating for increased durability and improved vision
- Combines well with 3M™ Aura™ 9300+ Series Disposable Respirators
- Safety glasses designed with user comfort and convenience in mind
- Strong polycarbonate lens for impact resistance
- Lightweight (26g) for maximum comfort all day
- Integrated browguard for increased protection
- Soft inner padding on temple for increased comfort and stability when worn
- Adjustable temple lengths in three positions for optimal comfort and fit
- Inclinable lens for easy adjustment and maximum comfort
Specifications
Specifications
| Certifications/Compliance | CE Approved, EN 166:2001 |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Amber |
| Product Type | Safety glasses |
| Frame Color | Dark Gray |
| Lens Material | Polycarbonate |