Learn More
Epredia™ Anti-Roll Plates for Epredia Cryostats
Product Code. 12093049 Shop All Epredia Products
Change view
Click to view available options
Diameter (Metric):
39.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12093049 39.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12093049 Supplier Epredia™ Supplier No. 449990
  • ONLINE EXCLUSIVE PRICE
    687.00 NOK
    584.00 NOK / Pack of 1
    Save 103.00 NOK
    14% Off
View all promotions
In Stock
Add to basket
Edge
Specifications
Product Type Anti Roll Plate
For Use With (Equipment) HM 525 NX with EC Blade Holder
Material Plastic
Diameter (Metric) 39.5 mm