missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen A24550
Additional Details : Weight : 0.01000kg
Description
Rat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Cross-Adsorbed in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
FITC | |
Goat | |
Affinity Chromatography | |
RUO | |
Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |