Learn More
anti-PTGS1, Human, IP-WB Antibody Pair, Abnova™
Product Code. 16048498
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16048498 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16048498 Supplier Abnova Supplier No. H00005742PW1.1Set
  • ONLINE EXCLUSIVE PRICE
    11560.00 NOK
    11089.94 NOK / 1 sets
    Save 470.06 NOK
    4% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Includes
Antibody pair for IP: rabbit polyclonal anti-PTGS1 (300μL), Antibody pair for WB: mouse purified polyclonal anti-PTGS1 (50μg)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen PTGS1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene PTGS1
Gene Alias COX1/COX3/PCOX1/PGG/HS/PGHS-1/PGHS1/PHS1/PTGHS
Gene Symbols PTGS1
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5742
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PTGS1 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-PTGS1 (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less
For Research Use Only