missing translation for 'onlineSavingsMsg'
Learn More

p54nrb Mouse, Unlabeled, Clone: 3, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611278

 View more versions of this product

Product Code. 15855859

  • ONLINE EXCLUSIVE PRICE
    2890.00 NOK
    2885.79 NOK / 1 set
    Save 4.21 NOK
    0.15% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Splicing, the removal of introns from pre-mRNA, is mediated by spliceosomal complexes that contain multiple snRNPs and a number of non-snRNP splicing factors, such as hPRP17 and hPRP18. p54 [nrb] (nuclear RNA-binding protein, 54 kDa) is a non-snRNP RNA splicing factor. It binds and enhances the activity of topoisomerase I, the enzyme that nicks DNA to relieve torsional stress during replication and transcription. p54 [nrb] is the the human homolog of mouse NonO and bovine IPEB. The N-terminal half of p54 [nrb] contains two RNA recognition motifs (RRMs), that allow it to bind single stranded RNA. These motifs are included in a region that is 71% homologous with the human splicing factor PSF and 42% homologous with the Drosophila puff-specific protein BJ6. In addition to its role in RNA splicing, p54 [nrb] functions as a DNA transcription factor in multiple cell types. The N-terminal portion of p54 [nrb] also mediates its interaction with DNA. The ability of p54 [nrb] to bind DNA, but not RNA, is thought to require post-translational modification. Thus, p54 [nrb] can function as either an RNA splicing factor or a DNA transcription factor in a post-translational-dependent manner.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

p54nrb
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human p54 [nrb] aa. 368-471
50 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Western Blot
3
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.