Learn More
NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human NDUFS6 protein.
Brand: Abnova H00004726-B01P.50ug
Description
The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria. The iron-sulfur protein (IP) fraction is made up of 7 subunits, including NDUFS6. See NDUFS1 (MIM 157655).[supplied by OMIM
Sequence: MAAAMTFCRLLNRCGEAARSLPLGARCFGVRVSPTGEKVTHTGQVYDDKDYRRIRFVGRQKEVNENFAIDLIAEQPVSEVETRVIACDGGGGALGHPKVYINLDKETKTGTCGYCGLQFRQHHHSpecifications
| NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human NDUFS6 protein. | |
| NDUFS6 | |
| NDUFS6 | |
| NDUFS6 (NP_004544.1, 1 a.a. ∼ 124 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004553.3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 4726 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.